Learn More
Grainger ELGA Hygienic Sample Point, Stainless Steel, 1 ea
Catalog No. 502788236
Encompass_Preferred
This hygienic sample point is designed to allow for the collection of representative water samples from your purification system. It is suitable for use in pharmaceutical, laboratory, and industrial water systems to ensure consistent monitoring and quality control.
  • Constructed from high-quality stainless steel
  • Designed for integration with compatible water purification systems
  • Enables reliable and hygienic water sampling
  • Compatible with MB007BPM1 and MB015BPM1 system models
Catalog No. 50-278-8236 Supplier Grainger Supplier No. 16D234
This hygienic sample point is designed to allow for the collection of representative water samples from your purification system. It is suitable for use in pharmaceutical, laboratory, and industrial water systems to ensure consistent monitoring and quality control.
  • Constructed from high-quality stainless steel
  • Designed for integration with compatible water purification systems
  • Enables reliable and hygienic water sampling
  • Compatible with MB007BPM1 and MB015BPM1 system models