Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hygric acid (N-Methyl-L-proline) | 475-11-6 | 98.0% | 129.16 | 250 MG
SDP

Catalog No. 5000402477
Encompass_Preferred

Hygric acid (N-Methyl-L-proline) is a proline analogue found in citrus juices and bergamot juice.

  • Purity: 98.0%
  • Synonyms: N-Methyl-L-proline
  • Molecular weight: 129.16
  • Chemical formula: C6H11NO2
  • Appearance: Solid
  • Color: White to off-white
  • High solubility in water and DMSO (100 mg/mL)
  • Recommended storage for powder: -20°C for 3 years, 4°C for 2 years
  • Recommended storage in solvent: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-02477 Supplier Medchemexpress LLC Supplier No. 250MGHY21754
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hygric acid (N-Methyl-L-proline) is a proline analogue found in citrus juices and bergamot juice.

  • Purity: 98.0%
  • Synonyms: N-Methyl-L-proline
  • Molecular weight: 129.16
  • Chemical formula: C6H11NO2
  • Appearance: Solid
  • Color: White to off-white
  • High solubility in water and DMSO (100 mg/mL)
  • Recommended storage for powder: -20°C for 3 years, 4°C for 2 years
  • Recommended storage in solvent: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.