Learn More
Thermco Products Inc HYGRO/THERMOMETER W/ALARM 1/EA
Catalog No. NC1211977
Encompass_Preferred
Thermo/Hygrometer with Hi-Low audible and visual alarm for all parameters. Temp -40� to 70�C ( -40� to 158�F). Dew Point Range: -40� to 70�C ( -40� to 158�F). Humidity Range: 0 to 99% RH. Min/max temperature and humidity memory recalling.
Catalog No. NC1211977 Supplier Thermco Products Inc Supplier No. ACCDTA100
Thermo/Hygrometer with Hi-Low audible and visual alarm for all parameters. Temp -40� to 70�C ( -40� to 158�F). Dew Point Range: -40� to 70�C ( -40� to 158�F). Humidity Range: 0 to 99% RH. Min/max temperature and humidity memory recalling.