Learn More
Cole-Parmer Extech 445814 Big Digit Thermohygrometer with Dew Point Point and Alarm
Catalog No. 50991911 Shop All Cole-Parmer Products
Encompass
Designed to monitor relative humidity, and temperature. Features include a super large display, C/F switchable, Min/Max with reset function, low battery indication and a built-in stand for sitting on a desk and hardware for mounting on a wall.
Catalog No. 50-991-911 Supplier Cole-Parmer Supplier No. 3780917
Designed to monitor relative humidity, and temperature. Features include a super large display, C/F switchable, Min/Max with reset function, low battery indication and a built-in stand for sitting on a desk and hardware for mounting on a wall.