Learn More
Hygro99, G-Biosciences

Description
It is over 99% pure and should be used for the most sensitive applications and on the most sensitive cell lines. All impurities that might cause inhibition of resistant cell lines have been removed.
Specifications
Specifications
| Form | Powder |
| Product Line | Gibco |
| Purity or Quality Grade | Laboratory |
| Quantity | 1 g |
| Molecular Weight (g/mol) | 563.52 |
| Purity | >99% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.