Learn More
Invivogen Hygromycin B Gold, 1g
SDP
Catalog No. NC9163750
Encompass
HygroGold, CAS No.: 31282-04-9, Molecular Formula: C20H37N3O13 HCl, Molecular Weight: 563.5, 1g liquid at 100mg/mL, Pack: 5 x 2mL/TB
Catalog No. NC9163750 Supplier Invivogen Supplier No. ANTHG1
HygroGold, CAS No.: 31282-04-9, Molecular Formula: C20H37N3O13 HCl, Molecular Weight: 563.5, 1g liquid at 100mg/mL, Pack: 5 x 2mL/TB