Learn More
GENIE GROUP Thermometer; Big Digit Hygro-; -10 to 60 deg C; With Mounting Bracket; Sensor
Catalog No. NC1503489
Encompass_Preferred
Max/Min with reset functionRear calibration adjustment potHumidity: 10 to 99% RHTemperature: 14 to 140°F (-10 to 60°C)Optional RH calibration bottlesAccuracy: ±4%RH; ±1°C/1.8°FDimensions: 4.3 x 3.9 x 0.78" (109 x 99 x 20mm)Weight: 6oz (169g)Complete with built-in stand, mounting bracket, sensor, and 1 AAA battery
Catalog No. NC1503489 Supplier GENIE GROUP Supplier No. 445715NIST
Max/Min with reset functionRear calibration adjustment potHumidity: 10 to 99% RHTemperature: 14 to 140°F (-10 to 60°C)Optional RH calibration bottlesAccuracy: ±4%RH; ±1°C/1.8°FDimensions: 4.3 x 3.9 x 0.78" (109 x 99 x 20mm)Weight: 6oz (169g)Complete with built-in stand, mounting bracket, sensor, and 1 AAA battery