Learn More
Medchemexpress LLC Hygromycin A | 6379-56-2 | 511.48 | 1 MG
SDP
Catalog No. 5000414557
Encompass_Preferred
Hygromycin A is a Borrelia burgdorferi-selective antibiotic. It is a spirochete-specific antibacterial that is conducive to gut health and can be used for Lyme disease research.
  • Spirochete-specific antibacterial
  • Contributes to gut health
  • Used for Lyme disease research
  • Selectively kills Borrelia burgdorferi in vivo
  • Positively impacts gut microbiota composition, leading to eubiosis
Catalog No. 50-004-14557 Supplier Medchemexpress LLC Supplier No. HYN106171MG
Hygromycin A is a Borrelia burgdorferi-selective antibiotic. It is a spirochete-specific antibacterial that is conducive to gut health and can be used for Lyme disease research.
  • Spirochete-specific antibacterial
  • Contributes to gut health
  • Used for Lyme disease research
  • Selectively kills Borrelia burgdorferi in vivo
  • Positively impacts gut microbiota composition, leading to eubiosis