Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hygromycin A | 6379-56-2 | 511.48 | 10 MM/1 ML
SDP

Catalog No. 5000415963
Encompass_Preferred

Hygromycin A is a *Borrelia burgdorferi*-selective antibiotic. It is a spirochete-specific antibacterial that contributes to gut health and can be utilized for Lyme disease research.

  • *Borrelia burgdorferi*-selective antibiotic
  • Spirochete-specific antibacterial that promotes gut health
  • Can be used for Lyme disease research
  • Purity of 96.78%
  • Molecular weight: 511.48
  • Formula: C23H29NO12
  • CAS No.: 6379-56-2
  • Appearance: Solid
  • Color: White to light yellow
  • Structure classification: Antibiotics
  • Initial source: Microorganisms (*Streptomyces hygroscopicus*)

Catalog No. 50-004-15963 Supplier Medchemexpress LLC Supplier No. HYN1061710MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hygromycin A is a *Borrelia burgdorferi*-selective antibiotic. It is a spirochete-specific antibacterial that contributes to gut health and can be utilized for Lyme disease research.

  • *Borrelia burgdorferi*-selective antibiotic
  • Spirochete-specific antibacterial that promotes gut health
  • Can be used for Lyme disease research
  • Purity of 96.78%
  • Molecular weight: 511.48
  • Formula: C23H29NO12
  • CAS No.: 6379-56-2
  • Appearance: Solid
  • Color: White to light yellow
  • Structure classification: Antibiotics
  • Initial source: Microorganisms (*Streptomyces hygroscopicus*)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.