Learn More
Medchemexpress LLC Hygromycin A | 6379-56-2 | 511.48 | 25 MG
SDP
Catalog No. 5000390302
Encompass_Preferred
Hygromycin A is a *Borrelia burgdorferi*-selective antibiotic that is also a spirochete-specific antibacterial. It contributes to gut health and is utilized in Lyme disease research. In vivo, Hygromycin A selectively eliminates *Borrelia burgdorferi* and positively influences gut microbiota composition, promoting eubiosis.
  • Selective antibiotic for *Borrelia burgdorferi*
  • Spirochete-specific antibacterial
  • Contributes to gut health
  • Used for Lyme disease research
  • Promotes eubiosis by impacting gut microbiota
Catalog No. 50-003-90302 Supplier Medchemexpress LLC Supplier No. 25MGHYN10617
Hygromycin A is a *Borrelia burgdorferi*-selective antibiotic that is also a spirochete-specific antibacterial. It contributes to gut health and is utilized in Lyme disease research. In vivo, Hygromycin A selectively eliminates *Borrelia burgdorferi* and positively influences gut microbiota composition, promoting eubiosis.
  • Selective antibiotic for *Borrelia burgdorferi*
  • Spirochete-specific antibacterial
  • Contributes to gut health
  • Used for Lyme disease research
  • Promotes eubiosis by impacting gut microbiota