Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hygromycin A | 6379-56-2 | 511.48 | 5 MG
SDP

Catalog No. 5000414770
Encompass_Preferred

Hygromycin A is a Borrelia burgdorferi-selective antibiotic. It is a spirochete-specific antibacterial that contributes to gut health and can be used for Lyme disease research. Hygromycin A selectively kills Borrelia burgdorferi and positively impacts gut microbiota composition, leading to eubiosis.

  • Appearance: Solid
  • Color: White to light yellow
  • Initial source: Microorganisms (Streptomyces hygroscopicus)
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: 4°C, protect from light. In solvent: -80°C, 6 months; -20°C, 1 month (protect from light).

Catalog No. 50-004-14770 Supplier Medchemexpress LLC Supplier No. HYN106175MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hygromycin A is a Borrelia burgdorferi-selective antibiotic. It is a spirochete-specific antibacterial that contributes to gut health and can be used for Lyme disease research. Hygromycin A selectively kills Borrelia burgdorferi and positively impacts gut microbiota composition, leading to eubiosis.

  • Appearance: Solid
  • Color: White to light yellow
  • Initial source: Microorganisms (Streptomyces hygroscopicus)
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: 4°C, protect from light. In solvent: -80°C, 6 months; -20°C, 1 month (protect from light).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.