Learn More
Medchemexpress LLC Hygromycin A | 6379-56-2 | 511.48 | 50 MG
SDP
Catalog No. 5000416554
Encompass_Preferred
Hygromycin A is a selective antibiotic targeting *Borrelia burgdorferi*, acting as a spirochete-specific antibacterial that contributes to gut health. It is valuable in Lyme disease research, with in vivo studies showing its ability to selectively eradicate *Borrelia burgdorferi* and positively influence gut microbiota composition, leading to eubiosis.
  • Borrelia burgdorferi-selective antibiotic
  • Spirochete-specific antibacterial
  • Contributes to gut health
  • Useful in Lyme disease research
  • Selectively eradicates Borrelia burgdorferi in vivo
  • Positively impacts gut microbiota composition
Catalog No. 50-004-16554 Supplier Medchemexpress LLC Supplier No. HYN1061750MG
Hygromycin A is a selective antibiotic targeting *Borrelia burgdorferi*, acting as a spirochete-specific antibacterial that contributes to gut health. It is valuable in Lyme disease research, with in vivo studies showing its ability to selectively eradicate *Borrelia burgdorferi* and positively influence gut microbiota composition, leading to eubiosis.
  • Borrelia burgdorferi-selective antibiotic
  • Spirochete-specific antibacterial
  • Contributes to gut health
  • Useful in Lyme disease research
  • Selectively eradicates Borrelia burgdorferi in vivo
  • Positively impacts gut microbiota composition