Learn More
FUJIFILM Biosciences Hygromycin B
Catalog No. 5000450283
Encompass_Preferred
Hygromycin B is an aminoglycoside antibiotic active against prokaryotic and eukaryotic cells commonly used as a selection agent in cell culture For research use only RUO
Catalog No. 50-004-50283 Supplier FUJIFILM Biosciences Supplier No. 08906151
Hygromycin B is an aminoglycoside antibiotic active against prokaryotic and eukaryotic cells commonly used as a selection agent in cell culture For research use only RUO