Learn More
Enzo Life Sciences Hygromycin B (sterile-filtered solution in PBS), (1g), CAS Number: 31282-04-9
SDP
Catalog No. NC0807148
Encompass_Preferred
Protein synthesis inhibitor. Antibiotic. Inhibits growth of prokaryotic and eukaryotic microorganisms and mammalian cells. Inhibits protein synthesis at the translocation step on the 70S ribosome and causes misreading of the mRNA. Penetrates cells that have been permeabilized by virus infection, hence it can act as an effective antiviral agent. Alternative Name: O-6-Amino-6-deoxy-L-glycero-D-galacto-heptopyranosylidene-(1-2-3)-O-beta-D-talopyranosyl(1-5)-2-deoxy-N3-methyl-D-streptamine. Formula: C20H37N3O13. MW: 527.5. CAS Number: 31282-04-9. MI: 14: 4852. Purity: ≥90% (HPLC). Appearance: Liquid. In PBS. Use/Stability: Working hygromycin solutions (≤2mg/ml) are stable for up to 1 month when stored at +4°C. Long Term Storage: +4°C
Catalog No. NC0807148 Supplier Enzo Life Sciences Supplier No. ALX380306G001
Protein synthesis inhibitor. Antibiotic. Inhibits growth of prokaryotic and eukaryotic microorganisms and mammalian cells. Inhibits protein synthesis at the translocation step on the 70S ribosome and causes misreading of the mRNA. Penetrates cells that have been permeabilized by virus infection, hence it can act as an effective antiviral agent. Alternative Name: O-6-Amino-6-deoxy-L-glycero-D-galacto-heptopyranosylidene-(1-2-3)-O-beta-D-talopyranosyl(1-5)-2-deoxy-N3-methyl-D-streptamine. Formula: C20H37N3O13. MW: 527.5. CAS Number: 31282-04-9. MI: 14: 4852. Purity: ≥90% (HPLC). Appearance: Liquid. In PBS. Use/Stability: Working hygromycin solutions (≤2mg/ml) are stable for up to 1 month when stored at +4°C. Long Term Storage: +4°C