Learn More
Bioworld Hygromycin B, 1 g
Catalog No. NC9719274
Encompass_Preferred
Hygromycin B is an antibiotic produced by S. hygroscopicus and is an aminoglycoside that kills bacteria, fungi, and higher eukaryotic cells by inhibition of their protein synthesis. It stabilizes the tRNA-ribosomal acceptor site which inhibits translation. It is also active against prokaryotic cells and is used in the lab for selection and maintenance of cells that contain the hygromycin resistance gene.Synonym: Hygrovetine; Hydromycin BPurity: >95% min (HPLC)Potency 1100 µg/mgHeavy Metals as Pb: <20 ppmCytotoxicity: Positive (CHO cell line)Endotoxicity: <10 EU/mgFor in vitro and lab research use only, not intended for human use or consumption. MUST SHIP COLD.Appearance: SolidRTECS Number: WK2130000
Catalog No. NC9719274 Supplier Bioworld Supplier No. 408100004
Hygromycin B is an antibiotic produced by S. hygroscopicus and is an aminoglycoside that kills bacteria, fungi, and higher eukaryotic cells by inhibition of their protein synthesis. It stabilizes the tRNA-ribosomal acceptor site which inhibits translation. It is also active against prokaryotic cells and is used in the lab for selection and maintenance of cells that contain the hygromycin resistance gene.Synonym: Hygrovetine; Hydromycin BPurity: >95% min (HPLC)Potency 1100 µg/mgHeavy Metals as Pb: <20 ppmCytotoxicity: Positive (CHO cell line)Endotoxicity: <10 EU/mgFor in vitro and lab research use only, not intended for human use or consumption. MUST SHIP COLD.Appearance: SolidRTECS Number: WK2130000