Learn More
Medchemexpress LLC HY-B0490 1g Medchemexpress, Hygromycin B CAS:31282-04-9 Purity:>98%
SDP
Catalog No. 502029399
Encompass_Preferred
Medchemexpress, HY-B0490 1g Hygromycin B CAS:31282-04-9 Hygromycin B is an aminoglycoside antibiotic active against prokaryotic and eukaryotic cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9399 Supplier Medchemexpress LLC Supplier No. HYB04901G
Medchemexpress, HY-B0490 1g Hygromycin B CAS:31282-04-9 Hygromycin B is an aminoglycoside antibiotic active against prokaryotic and eukaryotic cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.