Learn More
Selleck Chemical LLC Hygromycin B 250mg 31282-04-9
SDP
Catalog No. 501363187
Encompass_Preferred
Hygromycin B, a selective antibiotic that is effective on most bacteria, fungi and higher eukaryotes, inhibits protein synthesis by interfering with translocation and causing mistranslation at the 70S ribosome. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3187 Supplier Selleck Chemical LLC Supplier No. S2908250MG
Hygromycin B, a selective antibiotic that is effective on most bacteria, fungi and higher eukaryotes, inhibits protein synthesis by interfering with translocation and causing mistranslation at the 70S ribosome. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.