Learn More
Medchemexpress LLC Hygromycin B | 31282-04-9 | 99.9% | 1 ML
SDP
Catalog No. 5000407991
Encompass_Preferred
Hygromycin B is an aminoglycoside antibiotic active against both prokaryotic and eukaryotic cells. Synthesized by *Streptomyces hygroscopicus*, it functions by strongly inhibiting both 70S and 80S ribosomes, blocking polypeptide synthesis, and preventing ribosomal translocation. It is widely used in research for its ability to halt cell growth and confers resistance in transgenic organisms expressing the bacterial *hygR* gene.
  • Aminoglycoside antibiotic active against prokaryotic and eukaryotic cells
  • Inhibits both 70S and 80S ribosomes
  • Blocks polypeptide synthesis and prevents ribosomal translocation
  • Derived from *Streptomyces hygroscopicus*
  • Useful for halting yeast cell growth in research
  • Confers resistance to organisms expressing the bacterial *hygR* gene
Catalog No. 50-004-07991 Supplier Medchemexpress LLC Supplier No. HYB049010MM/1ML
Hygromycin B is an aminoglycoside antibiotic active against both prokaryotic and eukaryotic cells. Synthesized by *Streptomyces hygroscopicus*, it functions by strongly inhibiting both 70S and 80S ribosomes, blocking polypeptide synthesis, and preventing ribosomal translocation. It is widely used in research for its ability to halt cell growth and confers resistance in transgenic organisms expressing the bacterial *hygR* gene.
  • Aminoglycoside antibiotic active against prokaryotic and eukaryotic cells
  • Inhibits both 70S and 80S ribosomes
  • Blocks polypeptide synthesis and prevents ribosomal translocation
  • Derived from *Streptomyces hygroscopicus*
  • Useful for halting yeast cell growth in research
  • Confers resistance to organisms expressing the bacterial *hygR* gene