Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hygromycin B | 31282-04-9 | MFCD06795479 | >98.0% | 527.52 g/mol | C20H37N3O13 | 10 MG
SDP

Catalog No. 5000297104
Encompass_Preferred

Hygromycin B is an aminoglycoside antibiotic supplied as an analytical standard for research and analytical applications. It is active against prokaryotic and eukaryotic cells and is provided as a laboratory reagent for analytical testing (not for human therapeutic use).

  • Analytical standard for research and analytical applications.
  • Purity >98%.
  • Pack size 10 mg.
  • CAS number 31282-04-9.
  • Molecular formula C20H37N3O13 and molecular weight 527.52 g/mol.
  • Solubility: water 50 mg/mL (ultrasonic); DMSO 100 mg/mL (ultrasonic, warming to 60°C).
  • Provided as a solid powder suitable for analytical use.

Catalog No. 50-002-97104 Supplier Medchemexpress LLC Supplier No. HYB0490R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hygromycin B is an aminoglycoside antibiotic supplied as an analytical standard for research and analytical applications. It is active against prokaryotic and eukaryotic cells and is provided as a laboratory reagent for analytical testing (not for human therapeutic use).

  • Analytical standard for research and analytical applications.
  • Purity >98%.
  • Pack size 10 mg.
  • CAS number 31282-04-9.
  • Molecular formula C20H37N3O13 and molecular weight 527.52 g/mol.
  • Solubility: water 50 mg/mL (ultrasonic); DMSO 100 mg/mL (ultrasonic, warming to 60°C).
  • Provided as a solid powder suitable for analytical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.