Learn More
Medchemexpress LLC HY-B0490 5g , Hygromycin B Hygrovetine CAS:31282-04-9 Purity:98%
SDP
Catalog No. 501492182
Encompass_Preferred
Medchemexpress, HY-B0490 5g Hygromycin B Hygrovetine CAS:31282-04-9 Formula:C20H37N3O13 Target:Antibacterial; Antifungal Purity:98% Hygromycin B is an aminoglycoside antibiotic active against prokaryotic and eukaryotic cells. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2182 Supplier Medchemexpress LLC Supplier No. HYB04905G
Medchemexpress, HY-B0490 5g Hygromycin B Hygrovetine CAS:31282-04-9 Formula:C20H37N3O13 Target:Antibacterial; Antifungal Purity:98% Hygromycin B is an aminoglycoside antibiotic active against prokaryotic and eukaryotic cells. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.