Learn More
Gold Biotechnology Inc Hygromycin B 5 x 1 g
SDP
Supplier:  Gold Biotechnology Inc H27051
Encompass_Preferred
Hygromycin B is an aminoglycoside antibiotic isolated from Streptomyces hygroscopicus. It is commonly used to study protein synthesis. Aside from its efficacy against bacteria, fungi, and higher eukaryotic cells, hygromycin B is also an antiviral agent inhibiting translation upon selectively entering cells made permeable due to viral infection. It works as a standard selection antibiotic in gene experiments, especially for selection of hygromycin resistance gene transformed cells.
Catalog No. 50-153-2792