Learn More
Selleck Chemical LLC Hygromycin B S2908-1g
SDP
Catalog No. 503111685
Encompass_Preferred
Hygromycin B (Hygrovetine) a selective antibiotic that is effective on most bacteria fungi and higher eukaryotes inhibits protein synthesis by interfering with translocation and causing mistranslation at the 70S ribosome
Catalog No. 50-311-1685 Supplier Selleck Chemical LLC Supplier No. S29081G
Hygromycin B (Hygrovetine) a selective antibiotic that is effective on most bacteria fungi and higher eukaryotes inhibits protein synthesis by interfering with translocation and causing mistranslation at the 70S ribosome