Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Hygromycin B, Calbiochem™,

Catalog No. 4000511MU Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mU
10 mU
Packaging:
Alu drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
40-005-11MU 1 mU Alu drum
40-005-110MU 10 mU Alu drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 40-005-11MU Supplier MilliporeSigma™ Supplier No. 4000511MU

missing translation for 'validMainframeMsg'

Recommended Storage

2°C to 8°C, Aqueous stock solutions should be kept at concentrations of ≥50mg/ml and can be stored at either 4°C or −20°C Aqueous stock solutions (≥50mg/ml) are Stable for up to 6 months at 4°C or −20°C Working so

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31282-04-9
Molecular Formula C20H37N3O13
Molecular Weight (g/mol) 527.524
InChI Key GRRNUXAQVGOGFE-BBMONYMYSA-N
Synonym hygromycin b
PubChem CID 134129613
IUPAC Name (3'S,3aS,4S,4'S,5'R,6R,6'R,7R,7aS)-4-[(1R,2S,3R,5S,6S)-3-amino-2,6-dihydroxy-5-(methylamino)cyclohexyl]oxy-6'-[(1S)-1-amino-2-hydroxyethyl]-6-(hydroxymethyl)spiro[4,6,7,7a-tetrahydro-3aH-[1,3]dioxolo[4,5-c]pyran-2,2'-oxane]-3',4',5',7-tetrol
SMILES CNC1CC(C(C(C1O)OC2C3C(C(C(O2)CO)O)OC4(O3)C(C(C(C(O4)C(CO)N)O)O)O)O)N

Specifications

Color Brown
Packaging Alu drum
Quantity 1 mU
Physical Form Liquid
Recommended Storage 2°C to 8°C, Aqueous stock solutions should be kept at concentrations of ≥50mg/mL and can be stored at either 4°C or −20°C Aqueous stock solutions (≥50mg/mL) are Stable for up to 6 months at 4°C or −20°C. Working solutions (< 2 mg/ml) are stable for up to 1 month at 4°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.