Learn More
Cayman Chemical Hygromycin B-d4, Purity: ≥99% deuterated forms (d1-d4), 1 mg
SDP
Catalog No. 502949566
Encompass_Preferred
An internal standard for the quantification of hygromycin by GC- or LC-MS
Catalog No. 50-294-9566 Supplier Cayman Chemical Supplier No. 344431 MG
An internal standard for the quantification of hygromycin by GC- or LC-MS