Learn More
Cayman Chemical HygromycIn B-d4 5mg
SDP
Catalog No. 502948998
Encompass_Preferred
An internal standard for the quantification of hygromycin by GC- or LC-MS
Catalog No. 50-294-8998 Supplier Cayman Chemical Supplier No. 344435 MG
An internal standard for the quantification of hygromycin by GC- or LC-MS