Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hygromycin B, Fisher BioReagents

Catalog No. BP29521MU Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 mU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP29521MU 1 mU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP29521MU

missing translation for 'validMainframeMsg'

Inhibits protein synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31282-04-9 , 7732-18-5
Molecular Formula C20H37N3O13
Molecular Weight (g/mol) 527.524
InChI Key GRRNUXAQVGOGFE-BBMONYMYSA-N
PubChem CID 134129613
IUPAC Name (3'S,3aS,4S,4'S,5'R,6R,6'R,7R,7aS)-4-[(1R,2S,3R,5S,6S)-3-amino-2,6-dihydroxy-5-(methylamino)cyclohexyl]oxy-6'-[(1S)-1-amino-2-hydroxyethyl]-6-(hydroxymethyl)spiro[4,6,7,7a-tetrahydro-3aH-[1,3]dioxolo[4,5-c]pyran-2,2'-oxane]-3',4',5',7-tetrol
SMILES CNC1CC(C(C(C1O)OC2C3C(C(C(O2)CO)O)OC4(O3)C(C(C(C(O4)C(CO)N)O)O)O)O)N

Specifications

Type Hygromycin B
Physical Form Liquid
Source Streptomyces hygroscopicus
Packaging Glass Bottle
Sterile Filtered Pass Test
Color Yellow
Quantity 1 mU
Assay Percent Range > 80 %
Formula Weight 527.5
Percent Purity >80 %
Chemical Name or Material Hygromycin B
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
May be fatal if inhaled, absorbed through skin, or swallowed. Risk of serious damage to eyes. Lachrymator (substance which increases the flow of tears). May cause allergic respiratory and skin reaction. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.

NFPA

Health:4
Flammability:0
Instability:0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.