Learn More
Hygromycin B, G-Biosciences
SCI_ED
Catalog No. S139789
Change view
Click to view available options
Quantity:
25 g
Catalog No. Quantity
S139789 25 g
1 options
Catalog No. S139789 Supplier G-Biosciences Supplier No. RC860
Add to Cart
Add to Cart
Hygromycin B is produced by the bacterium Streptomyces hygroscopicus.
Hygromycin B is an aminocyclitol with a wide spectrum of activity that kills bacteria, fungi and higher eukaryotic cells by inhibiting protein synthesis.
Specifications
Form Powder
Product Type Antibiotic
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives None
Without Additives None
Quantity 25 g