Learn More
Enzo Life Sciences Hygromycin B (liquid) (1MU). CAS: 31282-04-9
SDP
Catalog No. 502008938
Encompass_Preferred
Antibiotic with high activity. Inhibits growth of prokaryotic and eukaryotic microorganisms and mammalian cells. Inhibits protein synthesis at the translocation step on the 70S ribosome and causes misreading of the mRNA. Penetrates cells that have been permeabilized by virus infection, hence it can act as an effective antiviral agent. Alternative name: O-6-Amino-6-deoxy-L-glycero-D-galacto-heptopyranosylidene-(1-2-3)-O-beta-D-talopyranosyl(1-5)-2-deoxy-N3-methyl-D-streptamine. Purity: ≥80% (HPLC). Appearance: Dark brown solution. Long Term Storage: +4°C.
Catalog No. 50-200-8938 Supplier Enzo Life Sciences Supplier No. ALX380059UM01
Antibiotic with high activity. Inhibits growth of prokaryotic and eukaryotic microorganisms and mammalian cells. Inhibits protein synthesis at the translocation step on the 70S ribosome and causes misreading of the mRNA. Penetrates cells that have been permeabilized by virus infection, hence it can act as an effective antiviral agent. Alternative name: O-6-Amino-6-deoxy-L-glycero-D-galacto-heptopyranosylidene-(1-2-3)-O-beta-D-talopyranosyl(1-5)-2-deoxy-N3-methyl-D-streptamine. Purity: ≥80% (HPLC). Appearance: Dark brown solution. Long Term Storage: +4°C.