Learn More
Alkali Scientific Hygromycin B, Powder Form, 1 Gram
SDP
Catalog No. 5000659625
Encompass_Preferred
Hygromycin B in powder form (1g) is an antibiotic used for cell culture selection in genetic research. This reagent is crucial for maintaining the selection pressure on transfected cells that have integrated the hygromycin resistance gene. Hygromycin B is effective in selecting cells that have been successfully genetically modified, making it an essential tool in gene expression, gene knockout, and drug discovery research. The 1g powder form is ideal for researchers who need to prepare custom solutions or require a cost-effective way to conduct multiple experiments involving mammalian and other eukaryotic cell lines.
Catalog No. 50-006-59625 Supplier Alkali Scientific Supplier No. HYG002CP
Hygromycin B in powder form (1g) is an antibiotic used for cell culture selection in genetic research. This reagent is crucial for maintaining the selection pressure on transfected cells that have integrated the hygromycin resistance gene. Hygromycin B is effective in selecting cells that have been successfully genetically modified, making it an essential tool in gene expression, gene knockout, and drug discovery research. The 1g powder form is ideal for researchers who need to prepare custom solutions or require a cost-effective way to conduct multiple experiments involving mammalian and other eukaryotic cell lines.