Learn More
Enzo Life Sciences Hygromycin B (sterile-filtered solution in water) (1g). CAS: 31282-04-9
SDP
Catalog No. 502009027
Encompass_Preferred
Antibiotic. Inhibits growth of prokaryotic and eukaryotic microorganisms and mammalian cells. Inhibits protein synthesis at the translocation step on the 70S ribosome and causes misreading of the mRNA. Penetrates cells that have been permeabilized by virus infection, hence it can act as an effective antiviral agent. Alternative name: O-6-Amino-6-deoxy-L-glycero-D-galacto-heptopyranosylidene-(1-2-3)-O-beta-D-talopyranosyl(1-5)-2-deoxy-N3-methyl-D-streptamine. Purity: ≥90% (HPLC). Long Term Storage: +4°C.
Catalog No. 50-200-9027 Supplier Enzo Life Sciences Supplier No. ALX380309G001
Antibiotic. Inhibits growth of prokaryotic and eukaryotic microorganisms and mammalian cells. Inhibits protein synthesis at the translocation step on the 70S ribosome and causes misreading of the mRNA. Penetrates cells that have been permeabilized by virus infection, hence it can act as an effective antiviral agent. Alternative name: O-6-Amino-6-deoxy-L-glycero-D-galacto-heptopyranosylidene-(1-2-3)-O-beta-D-talopyranosyl(1-5)-2-deoxy-N3-methyl-D-streptamine. Purity: ≥90% (HPLC). Long Term Storage: +4°C.