Learn More
Hygromycin B Solution, 50 mg/mL, G-Biosciences

Description
Hygromycin B is an aminocyclitol with a wide spectrum of activity that kills bacteria, fungi and higher eukaryotic cells by inhibiting protein synthesis.
Specifications
Specifications
| Concentration | 50 mg/mL |
| Form | Liquid |
| Product Line | Gibco |
| Purity or Quality Grade | Molecular Biology |
| Quantity | 50 mL |
| Molecular Weight (g/mol) | 527.6 |
| Purity | >90% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.