Learn More
Sigma Aldrich Fine Chemicals Biosciences Hygromycin B solution from Streptomyces hygroscopicus >=60% (HPLC), 45-60 mg/mL in H2O, gamma-irradiated | 31282-04-9 | MFCD06795479 | 250MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences H5527250MG
Encompass
Hygromycin B solution from Streptomyces hygroscopicus >=60% (HPLC), 45-60 mg/mL in H2O, gamma-irradiated | Purity: >=60% (HPLC) | Mol Wt: 527.52 | 31282-04-9 | MFCD06795479 | 250MG
Catalog No. 50-174-8170