Learn More
Sigma Aldrich Fine Chemicals Biosciences Hygromycin B solution from Streptomyces hygroscopicus >=60% (HPLC), 45-60 mg/mL in H2O | 31282-04-9 | MFCD06795479 | 1G
Catalog No. 501748161
Encompass
Hygromycin B solution from Streptomyces hygroscopicus >=60% (HPLC), 45-60 mg/mL in H2O | Purity: >=60% (HPLC) | Mol Wt: 527.52 | 31282-04-9 | MFCD06795479 | 1G
Catalog No. 50-174-8161 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. H06541G
Hygromycin B solution from Streptomyces hygroscopicus >=60% (HPLC), 45-60 mg/mL in H2O | Purity: >=60% (HPLC) | Mol Wt: 527.52 | 31282-04-9 | MFCD06795479 | 1G