Learn More
Medchemexpress LLC Hygromycin B (Standard) | 31282-04-9 | 99.9% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB0490R
Encompass_Preferred
Hygromycin B (Standard) is an analytical standard of Hygromycin B, primarily intended for research and analytical applications. This aminoglycoside antibiotic is active against both prokaryotic and eukaryotic cells.
  • Analytical standard for research and analytical applications
  • Active against prokaryotic and eukaryotic cells
  • Used as a reference standard in HPLC, GC, and MS experiments
Catalog No. 50-004-51026