Learn More
Cayman Chemical HygromycIn Bhydrate 500mg
SDP
Catalog No. 502954548
Encompass_Preferred
An aminoglycoside antibiotic; active against E. coli (MIC = 150 UG/ml); cytotoxic to both prokaryotic and eukaryotic cells via inhibition of protein synthesis; has commonly been used in molecular and cell biology applications to select for transformed cells expressing the E. coli hygromycin resistance genes hyg or hph
Catalog No. 50-295-4548 Supplier Cayman Chemical Supplier No. 14291500 MG
An aminoglycoside antibiotic; active against E. coli (MIC = 150 UG/ml); cytotoxic to both prokaryotic and eukaryotic cells via inhibition of protein synthesis; has commonly been used in molecular and cell biology applications to select for transformed cells expressing the E. coli hygromycin resistance genes hyg or hph