Learn More
Electron Microscopy Sciences HyhiTAG™ Removable Thermal Labels 1"
SDP
Supplier:  Electron Microscopy Sciences 7742613W
Encompass_Preferred
High temperature removable labels for the identification of various containers and surfaces intended for autoclaving, dry heat sterilization and gamma irradiations. Removability of the labels should be tested on substrates in end-user conditions.
Size: White
Color: 1"
Core: Yes
Labels across: 1
Catalog No. 50-193-0405