Learn More
Aviva Systems Biology Corp HYLS1 Antibody - middle region (ARP55449_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55449_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HYLS1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HYLS1. Alternate names: FLJ32915, HLSApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61509