Learn More
Antibody Research Corp HyMAX™ Hybridoma Fusion & Cloning Supplement 20x
Catalog No. NC0574465
Encompass
Description: HyMax™ Hybridoma Fusion and Cloning Supplement is a blended supplement media containing various cytokines, growth factors, hormones and vitamins specifically formulated for the survival and clonality of newly developing hyridoma. It is formulated to replace feeder cells during new hybridoma development. It can also be used for fast recovery of frozen cells from liquid nitrogen. It also found to stabilize antibody production by hybridoma, while untreated may lose IgG titer.
Catalog No. NC0574465 Supplier Antibody Research Corp Supplier No. 113004
Description: HyMax™ Hybridoma Fusion and Cloning Supplement is a blended supplement media containing various cytokines, growth factors, hormones and vitamins specifically formulated for the survival and clonality of newly developing hyridoma. It is formulated to replace feeder cells during new hybridoma development. It can also be used for fast recovery of frozen cells from liquid nitrogen. It also found to stabilize antibody production by hybridoma, while untreated may lose IgG titer.