Learn More
AdipoGen Hymenidin (1 mg)
SDP
Supplier:  AdipoGen AGCN20503M001
Encompass_Preferred
Chemical. CAS: 107019-95-4. Formula: C11H12BrN5O . H2O. MW: 310.2 . 18.0. Isolated from sponge Stylissa sp. Inhibitor of CDK5/p25 (IC50=4µM) and GSK-3beta (IC50=12µM). Potentially useful in neuronal diseases research. Antagonist of serotonergic receptors. Shown to reduce voltage-dependent calcium elevation. Moderate anticancer compound. Inhibits cell growth in a panel of cancer cell lines at low µM range. Antiprotozoal agent. Antibacterial. Plays a role in the sponge antibacterial defense.
Catalog No. 50-205-3686