Learn More
Bachem Hymenistatin I
SDP
Supplier:  Bachem 4026849.0001
Encompass_Preferred
Hymenistatin I, Cyclo(-Pro-Pro-Tyr-Val-Pro-Leu-Ile-Ile), 129536-23-8, C47H72N8O9, Mr 893.14, 1mg. HS-I. Hymenistatin I (HS-I) exerts an immunosuppressive effect in the humoral and cellular immune responses comparable with that of cyclosporin A (CsA). Comparison of the influence of HS-I and CsA on cytokine production suggests that the mechanisms of the interaction with the immunological system are substantially different for the two compounds tested.
Catalog No. 50-382-805