Learn More
Cayman Chemical HyocholIc AcId-d4 1mg
SDP
Catalog No. 502941530
Encompass_Preferred
An internal standard for the quantification of hyocholic acid by GC- or LC-MS
Catalog No. 50-294-1530 Supplier Cayman Chemical Supplier No. 270331 MG
An internal standard for the quantification of hyocholic acid by GC- or LC-MS