Learn More
Medchemexpress LLC Hyocholic Acid-d4 | 547-75-1 | 99% | 412.60 | 1 MG
SDP
Catalog No. 5000437297
Encompass_Preferred
Hyocholic Acid-d4 is the deuterium labeled Hyocholic Acid. It is for research use only and not sold to patients. This compound can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS applications
  • Deuteration may affect pharmacokinetic and metabolic profiles
Catalog No. 50-004-37297 Supplier Medchemexpress LLC Supplier No. HY121238S1MG
Hyocholic Acid-d4 is the deuterium labeled Hyocholic Acid. It is for research use only and not sold to patients. This compound can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS applications
  • Deuteration may affect pharmacokinetic and metabolic profiles