Learn More
Medchemexpress LLC HY-N0169 100mg Medchemexpress, Hyodeoxycholic acid CAS:83-49-8 Purity:>98%
SDP
Catalog No. 501933442
Encompass_Preferred
Medchemexpress, HY-N0169 100mg Hyodeoxycholic acid CAS:83-49-8 Hyodeoxycholic acid is a secondary bile acid formed in the small intestine by the gut flora, and acts as a TGR5 (GPCR19) agonist, with an EC 50 of 31.6 µM in CHO cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3442 Supplier Medchemexpress LLC Supplier No. HYN0169100MG
Medchemexpress, HY-N0169 100mg Hyodeoxycholic acid CAS:83-49-8 Hyodeoxycholic acid is a secondary bile acid formed in the small intestine by the gut flora, and acts as a TGR5 (GPCR19) agonist, with an EC 50 of 31.6 µM in CHO cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.