Learn More
Cayman Chemical Hyodeoxycholic Acid, Purity: ≥90%, CAS: 83-49-8, 5 g
SDP
Catalog No. 502984655
Encompass_Preferred
A secondary bile acid; decreases plasma VLDL and LDL cholesterol levels and reduces fasting glucose levels and atherosclerotic lesion size in LDL receptor knockout mice fed a Western diet at 1.25% in the diet; serum levels are increased in patients with Crohn’s disease or ulcerative colitis
Catalog No. 50-298-4655 Supplier Cayman Chemical Supplier No. 202945 G
A secondary bile acid; decreases plasma VLDL and LDL cholesterol levels and reduces fasting glucose levels and atherosclerotic lesion size in LDL receptor knockout mice fed a Western diet at 1.25% in the diet; serum levels are increased in patients with Crohn’s disease or ulcerative colitis