Learn More
Medchemexpress LLC Hyodeoxycholic acid (Standard) | 83-49-8 | MFCD00003681 | 99.6% | 392.57 | 25 MG
SDP
Catalog No. 5000429512
Encompass_Preferred
Hyodeoxycholic acid (Standard) is the analytical standard of Hyodeoxycholic acid, intended for research and analytical applications. It is a secondary bile acid formed in the small intestine by the gut flora, and acts as a TGR5 (GPCR19) agonist, with an EC50 of 31.6 μM in CHO cells.
  • Analytical standard
  • Reference standard supplied assay
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
  • Appearance: Solid
  • Color: White to off-white
  • Structure classification: Steroids
  • Initial source: Endogenous metabolite, human gut microbiota metabolites
  • Shipping: Room temperature in continental US; may vary elsewhere
Catalog No. 50-004-29512 Supplier Medchemexpress LLC Supplier No. 25MGHYN0169R
Add to Cart
Add to Cart
Hyodeoxycholic acid (Standard) is the analytical standard of Hyodeoxycholic acid, intended for research and analytical applications. It is a secondary bile acid formed in the small intestine by the gut flora, and acts as a TGR5 (GPCR19) agonist, with an EC50 of 31.6 μM in CHO cells.
  • Analytical standard
  • Reference standard supplied assay
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
  • Appearance: Solid
  • Color: White to off-white
  • Structure classification: Steroids
  • Initial source: Endogenous metabolite, human gut microbiota metabolites
  • Shipping: Room temperature in continental US; may vary elsewhere