Learn More
Apexbio Technology LLC Hyoscyamine(Synonyms: Levsin, Anaspaz, Daturine, Atropine sulfate (isomer), Hyoscyamin, Tropine tropate, (-)-Hyoscyamine), 10mM (in 1mL DMSO), CAS: 101-31-5.
SDP
Catalog No. 5000060167
Encompass_Preferred
Hyoscyamine (CAS 101-31-5) is a small molecule antagonist targeting acetylcholine receptors (AChR) Acting via inhibition of cholinergic signaling it exhibits a potent inhibitory activity with an IC50 of approximately 7 5 nM By blocking AChRs hyoscyamine modulates neurotransmission pathways involved in autonomic regulation Research applications include studies exploring cholinergic function investigations into parasympathetic nervous system disorders and testing therapeutic strategies involving ACh modulation
Catalog No. 50-000-60167 Supplier Apexbio Technology LLC Supplier No. B16055.1
Hyoscyamine (CAS 101-31-5) is a small molecule antagonist targeting acetylcholine receptors (AChR) Acting via inhibition of cholinergic signaling it exhibits a potent inhibitory activity with an IC50 of approximately 7 5 nM By blocking AChRs hyoscyamine modulates neurotransmission pathways involved in autonomic regulation Research applications include studies exploring cholinergic function investigations into parasympathetic nervous system disorders and testing therapeutic strategies involving ACh modulation