Learn More
Aviva Systems Biology Corp HYOU1 Antibody - middle region (ARP46716_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46716_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HYOU1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HYOU1. Alternate names: DKFZp686N08236, ORP150, Grp170, HSP12AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62956