Learn More
Apexbio Technology LLC Hyper Gibson Assembly Cloning Kit(Synonyms: Gibson Assembly Kit, Gibson Cloning Kit, Gibson Assembly Cloning Kit), 50rxns.
SDP
Catalog No. 5000061364
Encompass_Preferred
The Hyper Gibson Assembly Cloning Kit is based on homologous recombination enabling the precise assembly of DNA fragments into plasmid vectors without introducing extraneous nucleotide sequences In contrast to classical restriction enzyme ligase cloning methods this technique uses short homologous overlaps (15 25 bp) between PCR-amplified inserts and linearized plasmid ends to facilitate recombination The provided reagent mixture optimized with appropriate cofactors enhances recombinant efficiency tolerating minor impurities from PCR products Applications in biomedical research include rapid precise gene assembly and cloning in molecular biology workflows
Catalog No. 50-000-61364 Supplier Apexbio Technology LLC Supplier No. K166150
The Hyper Gibson Assembly Cloning Kit is based on homologous recombination enabling the precise assembly of DNA fragments into plasmid vectors without introducing extraneous nucleotide sequences In contrast to classical restriction enzyme ligase cloning methods this technique uses short homologous overlaps (15 25 bp) between PCR-amplified inserts and linearized plasmid ends to facilitate recombination The provided reagent mixture optimized with appropriate cofactors enhances recombinant efficiency tolerating minor impurities from PCR products Applications in biomedical research include rapid precise gene assembly and cloning in molecular biology workflows