Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hypaconine | 63238-68-6 | 469.57 | 5 MG
SDP

Catalog No. 5000416032
Encompass_Preferred

Hypaconine is a C19-diterpenoid alkaloid isolated from Aconitum and Delphinium spp. It exhibits strong cardiac activity and is for research use only.

  • Purity: 99.0%
  • Appearance: Solid, white to off-white
  • Molecular weight: 469.57
  • Molecular formula: C24H39NO8
  • CAS No.: 63238-68-6
  • For research use only

Catalog No. 50-004-16032 Supplier Medchemexpress LLC Supplier No. HYN19235MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hypaconine is a C19-diterpenoid alkaloid isolated from Aconitum and Delphinium spp. It exhibits strong cardiac activity and is for research use only.

  • Purity: 99.0%
  • Appearance: Solid, white to off-white
  • Molecular weight: 469.57
  • Molecular formula: C24H39NO8
  • CAS No.: 63238-68-6
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.