Learn More
Cayman Chemical Hypaconitine, Purity: ≥95%, CAS: 6900-87-4, 10 mg
SDP
Catalog No. 502989360
Encompass_Preferred
A diterpene alkaloid with diverse biological activities; inhibits nerve stimulation-evoked contractions in isolated mouse phrenic nerve-diaphragm muscle preparations (IC50 = 118 nM); prevents carrageenan-induced paw edema in mice at 0.025 mg/kg; reduces acetic acid-induced writhing in mice (ED50 = 0.1 mg/kg); toxic to mice (LD50 = 1.9 mg/kg, s.c.)
Catalog No. 50-298-9360 Supplier Cayman Chemical Supplier No. 2965610 MG
A diterpene alkaloid with diverse biological activities; inhibits nerve stimulation-evoked contractions in isolated mouse phrenic nerve-diaphragm muscle preparations (IC50 = 118 nM); prevents carrageenan-induced paw edema in mice at 0.025 mg/kg; reduces acetic acid-induced writhing in mice (ED50 = 0.1 mg/kg); toxic to mice (LD50 = 1.9 mg/kg, s.c.)